Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715215.3 | XP_002715261.1 | 4 - 5 | 928 | 71 | |
| XM_008264108.2 | XP_008262330.1 | 4 - 5 | 874 | 71 | ||
| XM_008264110.2 | XP_008262332.1 | 4 - 5 | 903 | 71 | ||
More Information
Set Membership: |
|

BLZF1
Genomic Map