Assay Details
| Gene Symbol: COPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723323.3 | XP_002723369.1 | 527 | 65 | ||
| XM_017339352.1 | XP_017194841.1 | 528 | 65 | |||
| XM_017339353.1 | XP_017194842.1 | 209 | 65 | |||
More Information
Set Membership: |
|

COPS3
Genomic Map