Assay Details
| Gene Symbol: MARCHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722015.3 | XP_002722061.1 | 137 | 82 | ||
| XM_008249134.2 | XP_008247356.1 | 140 | 82 | |||
| XM_017337971.1 | XP_017193460.1 | 145 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002722015.3, XM_008249134.2, XM_017337971.1 transcripts.
Set Membership: |
|

MARCHF2
Genomic Map