Assay Details
| Gene Symbol: MARCHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722015.3 | XP_002722061.1 | 771 | 66 | ||
| XM_008249134.2 | XP_008247356.1 | 798 | 66 | |||
| XM_008249135.2 | XP_008247357.1 | 682 | 66 | |||
| XM_008249136.2 | XP_008247358.1 | 770 | 66 | |||
More Information
Set Membership: |
|

MARCHF2
Genomic Map