Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719904.3 | XP_002719950.1 | 2 - 3 | 691 | 56 | |
| XM_008272599.2 | XP_008270821.1 | 2 - 3 | 691 | 56 | ||
| XM_008272600.2 | XP_008270822.1 | 2 - 3 | 1443 | 56 | ||
More Information
Set Membership: |
|

OTUD5
Genomic Map