Assay Details
| Gene Symbol: LZIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716159.3 | XP_002716205.1 | 4 - 5 | 730 | 93 | |
| XM_008265946.2 | XP_008264168.1 | 4 - 5 | 561 | 93 | ||
| XM_008265947.2 | XP_008264169.1 | 5 - 6 | 799 | 93 | ||
| XM_008265948.2 | XP_008264170.1 | 5 - 6 | 795 | 93 | ||
| XM_017346596.1 | XP_017202085.1 | 2 - 3 | 242 | 93 | ||
More Information
Set Membership: |
|

LZIC
Genomic Map