Assay Details
| Gene Symbol: ARHGAP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270292.2 | XP_008268514.1 | 7 - 8 | 1453 | 62 | |
| XM_008270293.2 | XP_008268515.1 | 8 - 9 | 1649 | 62 | ||
| XM_017348457.1 | XP_017203946.1 | 7 - 8 | 1455 | 62 | ||
More Information
Set Membership: |
|

ARHGAP19
Genomic Map