Assay Details
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263028.2 | XP_008261250.1 | 8 - 9 | 782 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPQ| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263028.2 | XP_008261250.1 | 8 - 9 | 782 | 78 | |
Set Membership: |
|