Assay Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719038.3 | XP_002719084.1 | 2 - 3 | 475 | 62 | |
| XM_008270682.2 | XP_008268904.1 | 2 - 3 | 475 | 62 | ||
| XM_017348852.1 | XP_017204341.1 | 2 - 3 | 475 | 62 | ||
| XM_017348853.1 | XP_017204342.1 | 2 - 3 | 475 | 62 | ||
| XM_017348854.1 | XP_017204343.1 | 2 - 3 | 475 | 62 | ||
More Information
Set Membership: |
|

PIGL
Genomic Map