Assay Details
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271548.2 | XP_008269770.1 | 23 | 6769 | 95 | |
XM_008271549.2 | XP_008269771.1 | 23 | 3499 | 95 | ||
XM_008271550.2 | XP_008269772.1 | 23 | 3496 | 95 | ||
XM_017349213.1 | XP_017204702.1 | 23 | 6772 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
