Assay Details
| Gene Symbol: LOC100339432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249079.2 | XP_008247301.1 | 86 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249079.2 transcripts.
Set Membership: |
|

LOC100339432
Genomic Map