Assay Details
| Gene Symbol: RAD21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710742.3 | XP_002710788.1 | 10 - 11 | 1368 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAD21| Gene Symbol: RAD21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710742.3 | XP_002710788.1 | 10 - 11 | 1368 | 65 | |
Set Membership: |
|