Assay Details
| Gene Symbol: LOC100340860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719155.3 | XP_002719201.1 | 4 - 5 | 921 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100340860| Gene Symbol: LOC100340860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719155.3 | XP_002719201.1 | 4 - 5 | 921 | 80 | |
Set Membership: |
|