Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718242.3 | XP_002718288.1 | 11 - 12 | 1403 | 62 | |
| XM_002718243.3 | XP_002718289.1 | 11 - 12 | 1428 | 62 | ||
| XM_017348292.1 | XP_017203781.1 | 10 - 11 | 1363 | 62 | ||
| XM_017348293.1 | XP_017203782.1 | 10 - 11 | 1276 | 62 | ||
| XM_017348294.1 | XP_017203783.1 | 11 - 12 | 1437 | 62 | ||
More Information
Set Membership: |
|

FRMD6
Genomic Map