Assay Details
| Gene Symbol: ITPRIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339966.1 | XP_017195455.1 | 1 - 2 | 97 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017339966.1 transcripts.
Set Membership: |
|

ITPRIPL1
Genomic Map