Assay Details
| Gene Symbol: ARHGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713258.3 | XP_002713304.1 | 5 - 6 | 697 | 61 | |
| XM_008260822.2 | XP_008259044.1 | 8 - 9 | 738 | 61 | ||
| XM_008260823.2 | XP_008259045.1 | 8 - 9 | 871 | 61 | ||
| XM_008260825.2 | XP_008259047.1 | 5 - 6 | 1159 | 61 | ||
| XM_008260826.2 | XP_008259048.1 | 6 - 7 | 638 | 61 | ||
More Information
Set Membership: |
|

ARHGEF3
Genomic Map