Assay Details
| Gene Symbol: NIPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718280.3 | XP_002718326.1 | 2 - 3 | 257 | 74 | |
| XM_008269730.2 | XP_008267952.1 | 4 - 5 | 681 | 74 | ||
| XM_008269731.2 | XP_008267953.1 | 4 - 5 | 685 | 74 | ||
| XM_008269732.2 | XP_008267954.1 | 3 - 4 | 559 | 74 | ||
| XM_008269733.2 | XP_008267955.1 | 4 - 5 | 520 | 74 | ||
| XM_008269734.2 | XP_008267956.1 | 4 - 5 | 636 | 74 | ||
| XM_008269736.2 | XP_008267958.1 | 2 - 3 | 422 | 74 | ||
| XM_008269737.2 | XP_008267959.1 | 4 - 5 | 857 | 74 | ||
| XM_008269742.2 | XP_008267964.1 | 2 - 3 | 373 | 74 | ||
| XM_008269744.2 | XP_008267966.1 | 2 - 3 | 313 | 74 | ||
| XM_008269745.2 | XP_008267967.1 | 2 - 3 | 296 | 74 | ||
| XM_017348346.1 | XP_017203835.1 | 3 - 4 | 495 | 74 | ||
More Information
Set Membership: |
|

NIPA2
Genomic Map