Assay Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717482.3 | XP_002717528.1 | 6 - 7 | 1088 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACBD3| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717482.3 | XP_002717528.1 | 6 - 7 | 1088 | 74 | |
Set Membership: |
|