Assay Details
Gene Symbol: SLITRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712999.3 | XP_002713045.1 | 1 - 2 | 542 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712999.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
