Assay Details
| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718680.3 | XP_002718726.1 | 2 - 3 | 1228 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDZD8| Gene Symbol: PDZD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718680.3 | XP_002718726.1 | 2 - 3 | 1228 | 97 | |
Set Membership: |
|