Assay Details
| Gene Symbol: LOC100343496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269863.2 | XP_008268085.1 | 25 - 26 | 2836 | 68 | |
| XM_008269864.2 | XP_008268086.1 | 25 - 26 | 2833 | 68 | ||
| XM_017348677.1 | XP_017204166.1 | 24 - 25 | 2728 | 68 | ||
More Information
Set Membership: |
|

LOC100343496
Genomic Map