Assay Details
| Gene Symbol: LOC100343496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269863.2 | XP_008268085.1 | 26 - 27 | 2895 | 84 | |
| XM_008269864.2 | XP_008268086.1 | 26 - 27 | 2892 | 84 | ||
| XM_008269866.2 | XP_008268088.1 | 26 - 27 | 2790 | 84 | ||
| XM_017348677.1 | XP_017204166.1 | 25 - 26 | 2787 | 84 | ||
More Information
Set Membership: |
|

LOC100343496
Genomic Map