Assay Details
| Gene Symbol: LOC100343496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269863.2 | XP_008268085.1 | 27 - 28 | 3114 | 59 | |
| XM_008269864.2 | XP_008268086.1 | 27 - 28 | 3111 | 59 | ||
| XM_008269865.2 | XP_008268087.1 | 26 - 27 | 3051 | 59 | ||
| XM_008269866.2 | XP_008268088.1 | 27 - 28 | 3009 | 59 | ||
| XM_008269867.2 | XP_008268089.1 | 25 - 26 | 2928 | 59 | ||
| XM_017348677.1 | XP_017204166.1 | 26 - 27 | 3006 | 59 | ||
More Information
Set Membership: |
|

LOC100343496
Genomic Map