Assay Details
| Gene Symbol: LOC100343496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269863.2 | XP_008268085.1 | 29 - 30 | 3899 | 100 | |
| XM_008269864.2 | XP_008268086.1 | 29 - 30 | 3896 | 100 | ||
| XM_008269865.2 | XP_008268087.1 | 28 - 29 | 3836 | 100 | ||
| XM_008269866.2 | XP_008268088.1 | 29 - 30 | 3794 | 100 | ||
| XM_008269867.2 | XP_008268089.1 | 27 - 28 | 3713 | 100 | ||
| XM_017348677.1 | XP_017204166.1 | 28 - 29 | 3791 | 100 | ||
More Information
Set Membership: |
|

LOC100343496
Genomic Map