Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710375.3 | XP_002710421.1 | 6 - 7 | 759 | 67 | |
| XM_008255379.2 | XP_008253601.1 | 7 - 8 | 840 | 67 | ||
| XM_017341251.1 | XP_017196740.1 | 6 - 7 | 758 | 67 | ||
| XM_017341252.1 | XP_017196741.1 | 6 - 7 | 757 | 67 | ||
More Information
Set Membership: |
|

GABRP
Genomic Map