Assay Details
| Gene Symbol: NT5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714557.3 | XP_002714603.1 | 6 - 7 | 1584 | 64 | |
| XM_017345271.1 | XP_017200760.1 | 6 - 7 | 1590 | 64 | ||
| XM_017345272.1 | XP_017200761.1 | 6 - 7 | 1590 | 64 | ||
More Information
Set Membership: |
|

NT5E
Genomic Map