Assay Details
| Gene Symbol: ACOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716151.3 | XP_002716197.2 | 6 - 7 | 723 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACOT7| Gene Symbol: ACOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716151.3 | XP_002716197.2 | 6 - 7 | 723 | 58 | |
Set Membership: |
|