Assay Details
| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270954.2 | XP_008269176.1 | 4 - 5 | 930 | 58 | |
| XM_008270955.2 | XP_008269177.1 | 4 - 5 | 938 | 58 | ||
| XM_008270956.2 | XP_008269178.1 | 4 - 5 | 931 | 58 | ||
| XM_008270959.2 | XP_008269181.1 | 4 - 5 | 935 | 58 | ||
| XM_017349146.1 | XP_017204635.1 | 4 - 5 | 1093 | 58 | ||
More Information
Set Membership: |
|

SLC6A4
Genomic Map