Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266633.2 | XP_008264855.1 | 3 | 4469 | 93 | |
| XM_008266634.2 | XP_008264856.1 | 3 | 4169 | 93 | ||
| XM_017346976.1 | XP_017202465.1 | 3 | 4322 | 93 | ||
| XM_017346977.1 | XP_017202466.1 | 4 | 4783 | 93 | ||
| XM_017346978.1 | XP_017202467.1 | 2 | 4214 | 93 | ||
More Information
Set Membership: |
|

B3GNT5
Genomic Map