Assay Details
| Gene Symbol: LOC100339446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708640.3 | XP_002708686.1 | 16 - 17 | 2171 | 89 | |
| XM_017344075.1 | XP_017199564.1 | 16 - 17 | 2172 | 89 | ||
| XM_017344083.1 | XP_017199572.1 | 16 - 17 | 1982 | 89 | ||
More Information
Set Membership: |
|

LOC100339446
Genomic Map