Assay Details
| Gene Symbol: LOC100339446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708640.3 | XP_002708686.1 | 9 - 10 | 1376 | 116 | |
| XM_017344075.1 | XP_017199564.1 | 9 - 10 | 1377 | 116 | ||
| XM_017344083.1 | XP_017199572.1 | 9 - 10 | 1187 | 116 | ||
More Information
Set Membership: |
|

LOC100339446
Genomic Map