Assay Details
| Gene Symbol: LCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711830.3 | XP_002711876.1 | 4 - 5 | 530 | 79 | |
| XM_017342533.1 | XP_017198022.1 | 4 - 5 | 532 | 79 | ||
| XM_017342534.1 | XP_017198023.1 | 3 - 4 | 439 | 79 | ||
More Information
Set Membership: |
|

LCMT1
Genomic Map