Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346564.1 | XP_017202053.1 | 18 - 19 | 4538 | 80 | |
| XM_017346566.1 | XP_017202055.1 | 15 - 16 | 4248 | 80 | ||
More Information
Set Membership: |
|

CRYBG2
Genomic Map