Assay Details
| Gene Symbol: TBC1D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714178.3 | XP_002714224.1 | 4 - 5 | 654 | 64 | |
| XM_002714179.3 | XP_002714225.1 | 5 - 6 | 804 | 64 | ||
| XM_008262399.2 | XP_008260621.1 | 5 - 6 | 734 | 64 | ||
| XM_008262401.2 | XP_008260623.1 | 5 - 6 | 714 | 64 | ||
More Information
Set Membership: |
|

TBC1D7
Genomic Map