Assay Details
| Gene Symbol: LMAN2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710023.3 | XP_002710069.1 | 8 - 9 | 970 | 64 | |
| XM_002710024.3 | XP_002710070.1 | 7 - 8 | 937 | 64 | ||
| XM_017339890.1 | XP_017195379.1 | 6 - 7 | 656 | 64 | ||
More Information
Set Membership: |
|

LMAN2L
Genomic Map