Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712358.3 | XP_002712404.1 | 5 - 6 | 939 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STK17B| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712358.3 | XP_002712404.1 | 5 - 6 | 939 | 67 | |
Set Membership: |
|