Assay Details
| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721037.3 | XP_002721083.1 | 820 | 75 | ||
| XM_008274340.2 | XP_008272562.1 | 826 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAAT5| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721037.3 | XP_002721083.1 | 820 | 75 | ||
| XM_008274340.2 | XP_008272562.1 | 826 | 75 | |||
Set Membership: |
|