Assay Details
Gene Symbol: SLC10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267641.2 | XP_008265863.1 | 6 - 7 | 1567 | 57 | |
XM_017347442.1 | XP_017202931.1 | 5 - 6 | 1467 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
