Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716275.3 | XP_002716321.1 | 2 - 3 | 439 | 110 | |
| XM_017346843.1 | XP_017202332.1 | 3 - 4 | 510 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716275.3, XM_017346843.1 transcripts.
Set Membership: |
|

GPR87
Genomic Map