Assay Details
| Gene Symbol: MBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718146.3 | XP_002718192.1 | 5 - 6 | 690 | 81 | |
| XM_008269505.2 | XP_008267727.1 | 5 - 6 | 691 | 81 | ||
| XM_008269506.2 | XP_008267728.1 | 4 - 5 | 467 | 81 | ||
More Information
Set Membership: |
|

MBIP
Genomic Map