Assay Details
| Gene Symbol: MED8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265410.1 | XP_008263632.1 | 2 - 3 | 205 | 70 | |
| XM_017346387.1 | XP_017201876.1 | 1 - 2 | 334 | 70 | ||
More Information
Set Membership: |
|

MED8
Genomic Map