Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708968.2 | XP_002709014.1 | 3 - 4 | 481 | 63 | |
| XM_008268758.2 | XP_008266980.1 | 3 - 4 | 454 | 63 | ||
| XM_008268768.2 | XP_008266990.1 | 3 - 4 | 397 | 63 | ||
| XM_017345548.1 | XP_017201037.1 | 3 - 4 | 439 | 63 | ||
More Information
Set Membership: |
|

HTATIP2
Genomic Map