Assay Details
| Gene Symbol: ZBTB22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714465.3 | XP_002714511.1 | 1 - 2 | 103 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002714465.3 transcripts.
Set Membership: |
|

ZBTB22
Genomic Map