Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262162.2 | XP_008260384.1 | 5 - 6 | 1173 | 122 | |
| XM_008262163.2 | XP_008260385.1 | 5 - 6 | 1291 | 122 | ||
| XM_008262164.2 | XP_008260386.1 | 5 - 6 | 1404 | 122 | ||
| XM_008262165.2 | XP_008260387.1 | 5 - 6 | 1364 | 122 | ||
| XM_017344703.1 | XP_017200192.1 | 4 - 5 | 1531 | 122 | ||
| XM_017344704.1 | XP_017200193.1 | 4 - 5 | 1471 | 122 | ||
| XM_017344705.1 | XP_017200194.1 | 5 - 6 | 1427 | 122 | ||
| XM_017344706.1 | XP_017200195.1 | 3 - 4 | 1429 | 122 | ||
More Information
Set Membership: |
|

ZNF131
Genomic Map