Assay Details
| Gene Symbol: ODAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252414.2 | XP_008250636.1 | 2328 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ODAD1| Gene Symbol: ODAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252414.2 | XP_008250636.1 | 2328 | 72 | ||
Set Membership: |
|