Assay Details
| Gene Symbol: GLP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714692.3 | XP_002714738.1 | 12 - 13 | 1323 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLP1R| Gene Symbol: GLP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714692.3 | XP_002714738.1 | 12 - 13 | 1323 | 63 | |
Set Membership: |
|