Assay Details
| Gene Symbol: GNPNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718249.3 | XP_002718295.1 | 4 - 5 | 583 | 115 | |
| XM_017348306.1 | XP_017203795.1 | 5 - 6 | 719 | 115 | ||
More Information
Set Membership: |
|

GNPNAT1
Genomic Map