Assay Details
| Gene Symbol: TARBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711024.3 | XP_002711070.1 | 8 - 9 | 1242 | 54 | |
| XM_008256539.2 | XP_008254761.1 | 8 - 9 | 1004 | 54 | ||
More Information
Set Membership: |
|

TARBP2
Genomic Map