Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710472.3 | XP_002710518.1 | 11 - 12 | 1505 | 67 | |
| XM_017341296.1 | XP_017196785.1 | 10 - 11 | 1238 | 67 | ||
More Information
Set Membership: |
|

PXDNL
Genomic Map