Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710472.3 | XP_002710518.1 | 6 - 7 | 664 | 70 | |
| XM_017341296.1 | XP_017196785.1 | 6 - 7 | 664 | 70 | ||
More Information
Set Membership: |
|

PXDNL
Genomic Map